Skip to Content
Merck
All Photos(1)

Key Documents

682209

Sigma-Aldrich

2-Mesityl-5-methylimidazo[1,5-a]pyridinium chloride

97%

Synonym(s):

2-(2,4,6-Trimethylphenyl)-5-methylimidazo[1,5-a]pyridinim chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19ClN2
CAS Number:
Molecular Weight:
286.80
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

mp

>300 °C

SMILES string

[Cl-].Cc1cc(C)c(c(C)c1)-[n+]2cc3cccc(C)n3c2

InChI

1S/C17H19N2.ClH/c1-12-8-13(2)17(14(3)9-12)18-10-16-7-5-6-15(4)19(16)11-18;/h5-11H,1-4H3;1H/q+1;/p-1

InChI key

DJFXURXZOPNPGY-UHFFFAOYSA-M

Application

Reactant for gold cycloisomerization catalytic reactions
Stable N-heterocyclic carbene precursor.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Manuel Alcarazo et al.
Journal of the American Chemical Society, 127(10), 3290-3291 (2005-03-10)
The imidazo[1,5-a]pyridine skeleton provides a versatile platform for the generation of new types of stable N-heterocyclic carbenes. Rh(I) mono- (6) and biscarbenes (7) from imidazo[1,5-a]pyridin-3-ylidenes (ImPy) and derivatives such as 13 from a mesoionic carbene were synthesized and characterized.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service