Skip to Content
Merck
All Photos(1)

Key Documents

673404

Sigma-Aldrich

1,3-Dicyclohexylbenzimidazolium chloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27ClN2
CAS Number:
Molecular Weight:
318.88
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

solid

reaction suitability

reagent type: catalyst

mp

216-220 °C

SMILES string

[Cl-].C1CCC(CC1)n2c[n+](C3CCCCC3)c4ccccc24

InChI

1S/C19H27N2.ClH/c1-3-9-16(10-4-1)20-15-21(17-11-5-2-6-12-17)19-14-8-7-13-18(19)20;/h7-8,13-17H,1-6,9-12H2;1H/q+1;/p-1

InChI key

JACUDTKAZHCIPU-UHFFFAOYSA-M

Application

  • Catalyst in rhodium-N-heterocyclic carbene complex-catalyzed O-arylation of phenols with arylbromides
  • Reactant in preparation of Pd-PEPPSI complexes for catalysis of cross-coupling reactions

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Emerging class of privileged ligands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service