Skip to Content
Merck
All Photos(1)

Key Documents

655651

Sigma-Aldrich

Dopamine-1,1,2,2-d4 hydrochloride

98 atom % D

Synonym(s):

2-(3,4-Dihydroxyphenyl)ethyl-1,1,2,2-d4-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8D4H7O2N.HCl
CAS Number:
Molecular Weight:
193.66
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

mp

248-250 °C (lit.)

mass shift

M+4

SMILES string

Cl.[2H]C([2H])(N)C([2H])([2H])c1ccc(O)c(O)c1

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H/i3D2,4D2;

InChI key

CTENFNNZBMHDDG-URZLSVTISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kate Thomson et al.
Experimental eye research, 200, 108233-108233 (2020-09-14)
Animal models have demonstrated a link between dysregulation of the retinal dopamine system and the development of experimental myopia (short-sightedness). However, pharmacological investigations of dopamine in animal models rely heavily on intravitreal or systemic administration, which have several limitations for
Silvia Giatti et al.
Journal of medicinal chemistry, 64(8), 4553-4566 (2021-04-13)
Finasteride, a 5-alpha reductase (5α-R) inhibitor, is a widely used drug for treating androgen-dependent conditions. However, its use is associated with sexual, psychological, and physical complaints, suggesting that other mechanisms, in addition to 5α-R inhibition, may be involved. Here, a
Celia Herrera-Rincon et al.
NPJ Regenerative medicine, 5, 2-2 (2020-02-13)
Infections have numerous effects on the brain. However, possible roles of the brain in protecting against infection, and the developmental origin and role of brain signaling in immune response, are largely unknown. We exploited a unique Xenopus embryonic model to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service