Skip to Content
Merck
All Photos(1)

Key Documents

645087

Sigma-Aldrich

5-Phenylisoxazole-3-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO3
CAS Number:
Molecular Weight:
189.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

160-164 °C (lit.)

SMILES string

OC(=O)c1cc(on1)-c2ccccc2

InChI

1S/C10H7NO3/c12-10(13)8-6-9(14-11-8)7-4-2-1-3-5-7/h1-6H,(H,12,13)

InChI key

XJYOBHXWBRKOQO-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical studies on isoxazoles. II A new fluorimetric determination of 5-phenyl-3-isoxazolecarboxylic acid and its application to the determination of perisoxal in plasma.
T Umeda et al.
Chemical & pharmaceutical bulletin, 28(12), 3507-3513 (1980-12-01)
Annamaria Lilienkampf et al.
Journal of medicinal chemistry, 53(2), 678-688 (2009-12-17)
New antituberculosis (anti-TB) drugs are urgently needed to shorten the 6-12 month treatment regimen and especially to battle drug-resistant Mycobacterium tuberculosis (Mtb) strains. In this study, we have continued our efforts to develop isoxazole-based anti-TB compounds by applying rational drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service