616338
D-Glucose-d12
97 atom % D
Synonym(s):
Deuterated Glucose, Labeled Glucose, Dextrose-d12
About This Item
Recommended Products
isotopic purity
97 atom % D
Quality Level
Assay
≥99% (HPLC)
form
powder
optical activity
[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)
technique(s)
bio NMR: suitable
mp
150-152 °C (lit.)
mass shift
M+12
SMILES string
[2H]OC([2H])([2H])[C@@]1([2H])OC([2H])(O[2H])[C@]([2H])(O[2H])[C@@]([2H])(O[2H])[C@]1([2H])O[2H]
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i1D2,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D
InChI key
WQZGKKKJIJFFOK-QIRXRFCNSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.
Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.
Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.
Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service