Skip to Content
Merck
All Photos(1)

Documents

613533

Sigma-Aldrich

1,6-Diaminohexane-1,1,6,6-d4

98 atom % D

Synonym(s):

1,6-Hexane-1,1,6,6-d4-diamine, Hexamethylene-1,1,6,6-d4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCD2(CH2)4CD2NH2
CAS Number:
Molecular Weight:
120.23
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

bp

204-205 °C (lit.)

mp

42-45 °C (lit.)

mass shift

M+4

SMILES string

[2H]C([2H])(N)CCCCC([2H])([2H])N

InChI

1S/C6H16N2/c7-5-3-1-2-4-6-8/h1-8H2/i5D2,6D2

InChI key

NAQMVNRVTILPCV-NZLXMSDQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service