Skip to Content
Merck
All Photos(1)

Key Documents

601551

Sigma-Aldrich

Boron-10B

≥99% 10B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
10B
CAS Number:
Molecular Weight:
10.01
UNSPSC Code:
12141907

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥99% 10B

form

solid

mol wt

10.11 by atom % calculation

InChI

1S/B/i1-1

InChI key

ZOXJGFHDIHLPTG-BJUDXGSMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. In what form it is avilable ?

    1 answer
    1. This product is available in solid form.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service