Skip to Content
Merck
All Photos(3)

Key Documents

596280

Sigma-Aldrich

6-Bromo-3-pyridinecarboxaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrNO
CAS Number:
Molecular Weight:
186.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

solid

mp

104-110 °C (lit.)

functional group

aldehyde
bromo

SMILES string

[H]C(=O)c1ccc(Br)nc1

InChI

1S/C6H4BrNO/c7-6-2-1-5(4-9)3-8-6/h1-4H

InChI key

PVUKGNBRJFTFNJ-UHFFFAOYSA-N

Application

6-Bromo-3-pyridinecarboxaldehyde can be used as a starting material in the synthesis of 2-(3′,5′-bis(trifluoromethyl)biphenyl-4-yl)-3-(6-(3,5-bis(trifluoromethyl)phenyl) pyridin-3-yl)acrylonitrile (Py-CN-TFMBE), which can have many potential applications as a stimuli-responsive fluorescent organo-gelator.[1] It can also be used in the preparation of porous organic polymers, which can act like a fluorescent sensor for detecting palladium(II) and organic dyes.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"A Thermoreversible and Proton-Induced Gel- Sol Phase Transition with Remarkable Fluorescence Variation"
Chung WJ, et al.
Chemistry of Materials, 20(21), 6750- 6755 (2008)
"Facile synthesis of porous organic polymers for the absorption of Pd (II) ions and organic dye"
Luo Y, et al.
Royal Society of Chemistry Advances, 6(83), 79781-79791 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service