Skip to Content
Merck
All Photos(3)

Key Documents

569607

Sigma-Aldrich

5-Bromo-2-iodopyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrIN
CAS Number:
Molecular Weight:
283.89
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

113-117 °C (lit.)

functional group

bromo
iodo

SMILES string

Brc1ccc(I)nc1

InChI

1S/C5H3BrIN/c6-4-1-2-5(7)8-3-4/h1-3H

InChI key

HSNBRDZXJMPDGH-UHFFFAOYSA-N

Application

5-Bromo-2-iodopyridine may be used in the synthesis of the following:
  • 5-bromopyridyl-2-magnesium chloride via reaction with isopropylmagnesium chloride solution[1]
  • 5-bromo-2-pyridylzinc iodide via treatment with active zinc in tetrahydrofuran[2]
  • 5-bromo-2-(trifluoromethyl)pyridine via halogen/trifluoromethyl displacement reaction
  • (5-bromopyridin-2-yl)zinc(II) chloride via multi-step synthesis[3]
  • 5,5′-dibromo-2,2′-bipyridine via multi-step synthesis[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"5-Bromo-2-pyridylzinc reagent; direct preparation and its coupling reactions"
Rieke.DR, et al.
Tetrahedron Letters, 52(2), 244-247 (2011)
"Synthesis of 5-bromopyridyl-2-magnesium chloride and its application in the synthesis of functionalized pyridines"
Song JJ, et al.
Organic Letters, 6(26), 4905-4907 (2004)
"A functional triazine framework based on N-heterocyclic building blocks"
Hug S, et al.
Journal of Materials Chemistry, 22(28), 13956- 13964 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service