Skip to Content
Merck
All Photos(2)

Key Documents

541443

Sigma-Aldrich

Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene]

average Mn 40,000-70,000

Synonym(s):

MEH-PPV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C18H28O2)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

description

Band gap: 2.3 eV

Quality Level

mol wt

average Mn 40,000-70,000

fluorescence

λex 493 nm; λem 554 nm in toluene

Orbital energy

HOMO -5.3 eV 
LUMO -3 eV 

OLED Device Performance

ITO/PEDOT:PSS/MEH-PPV/Al

  • Color: orange-red
  • Max. Luminance: 220 Cd/m2
  • Max. EQE: 0.3 %
  • Turn-On Voltage: 3.8 V

ITO/PEDOT:PSS/MEH-PPV/PBO/Al
  • Color: orange-red
  • Max. Luminance: 970 Cd/m2
  • Max. EQE: 1.9 %
  • Turn-On Voltage: 2.9 V

ITO/PEDOT:PSS/MEH-PPV/PBZT/Al
  • Color: orange-red
  • Max. Luminance: 1400 Cd/m2
  • Max. EQE: 2.5 %
  • Turn-On Voltage: 2.8 V

OPV Device Performance

ITO/MEH-PPV/BBL/Al

  • Short-circuit current density (Jsc): 1.98 mA/cm2
  • Open-circuit voltage (Voc): 0.93 V
  • Fill Factor (FF): 0.47
  • Power Conversion Efficiency (PCE): 1.1 %

ITO/MEH-PPV:PC61BM(1:4)/Ca
  • Short-circuit current density (Jsc): 2 mA/cm2
  • Open-circuit voltage (Voc): 0.8 V
  • Fill Factor (FF): 0.25
  • Power Conversion Efficiency (PCE): 1.5 %

Mw/Mn

~6

storage temp.

2-8°C

SMILES string

O(CC(CCCC)CC)c1cc(c(cc1)OC)C=C

InChI

1S/C17H26O2/c1-5-8-9-14(6-2)13-19-16-10-11-17(18-4)15(7-3)12-16/h7,10-12,14H,3,5-6,8-9,13H2,1-2,4H3

InChI key

CTUSWJPWMNVXEE-UHFFFAOYSA-N

General description

Light-emitting conjugated polymer.
MEH-PPV is a conducting organic semiconductor which has low molecular weight and hydrophobic characteristics. It is a poly(phenylenevinylene) (PPV) derivative and a conjugating polymer with highest occupied molecular orbital (HOMO) below the fermi level of gold.[1][2][3]

Application

Conducting polymer in solar cells and carbon nanotube OLEDs. Useful in producing bright and efficient white polymeric light emitting diodes.
Filter before application.
MEH-PPV can form a nanocomposite with carbon nanotubes (CNTs) by spin coating that can be used for H2S gas sensors.[4] It can also be used as a conducting polymer which results in the formation of a low-cost dopant free hole transporting material (HTM) for the fabrication of perovskite solar cells with a power efficiency of 9.65%.[5] MEH-PPV can be blended with PCPDTBT for use as a sensitizer for dye sensitized photo sensors (DSPS).[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective metal-vapor deposition on solvent evaporated polymer surfaces.
Yamaguchi K and Tsujioka T
Thin Solid Films, 597, 220-225 (2015)
A gold/organic semiconductor diode for ppm-level humidity sensing.
Hossein-Babaei F and Shabani P
Sensors and Actuators B, Chemical, 205, 143-150 (2014)
Binary blend based dye sensitized photo sensor using PCPDTBT and MEH-PPV composite as a light sensitizer.
Qadir KW, et al.
Synthetic Metals, 210(1), 392-397 (2015)
Shen, F.; et al.
Semiconductor Science and Technology, 22, L16-L16 (2006)
Enhancement of MEH-PPV: CNT for H2S gas sensor.
Ibrahim IM, et al.
Journal of Physics. Conference Series, 1032(1), 012003-012003 (2018)

Articles

PCBM-based n-type semiconductors - Find p- and n-type organic semiconductors available with PCBM library & properties.

PCBM-based n-type semiconductors - Find p- and n-type organic semiconductors available with PCBM library & properties.

PCBM-based n-type semiconductors - Find p- and n-type organic semiconductors available with PCBM library & properties.

PCBM-based n-type semiconductors - Find p- and n-type organic semiconductors available with PCBM library & properties.

See All

Questions

1–6 of 6 Questions  
  1. do you have molecular weight characterization data for your Poly[2-methoxy-5-(2’-ethylhexyloxy)-1,4-phenylene vinylene] samples?

    1 answer
    1. As listed in the 'PROPERTIES' section, the average molecular weight of this product is 40,000-70,000.

      Helpful?

  2. What is the glass transition temperature (Tg) of Product 541443, Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene]?

    1 answer
    1. Unfortunately, the glass transition temperature (Tg) is not determined for this product.

      Helpful?

  3. What is the solubility of Product 541443, Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene]?

    1 answer
    1. Per our quality control testing, this product is soluble in toluene with heat and sonication at a concentration of 0.02 g/L.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. Does Product 541443, Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene], contain a dopant?

    1 answer
    1. This product does not contain a dopant.

      Helpful?

  6. Can Product 541443, Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene], efficiently convert light to electricity?

    1 answer
    1. The polyphenylenevinylene (PPV) polymers themselves do not convert light to electricity very efficiently.  They need to be blended with an electron transport material. The best material for this is a fullerene material called [6,6]-Phenyl C61 butyric acid methyl ester (PCBM, offered as #684430, 684449, 684457). There are several references that discuss polymer-fullerene heterojunctions.  One paper on this subject is Brabec, C.J.; et. al. Applied Physics Letters, 80, 1288 (2002).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service