513512
Copper(I) trifluoromethanesulfonate toluene complex
≥99.7% trace metals basis
Synonym(s):
CuOTf-toluene, Copper(I) triflate toluene complex, Cuprous trifluoromethanesulfonate toluene complex, Trifluoromethanesulfonic acid copper(I) salt toluene complex
About This Item
Recommended Products
Quality Level
Assay
≥99.7% trace metals basis
reaction suitability
core: copper
reagent type: catalyst
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
, Aligned
SMILES string
[Cu+].[Cu+].Cc1ccccc1.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
InChI
1S/C7H8.2CHF3O3S.2Cu/c1-7-5-3-2-4-6-7;2*2-1(3,4)8(5,6)7;;/h2-6H,1H3;2*(H,5,6,7);;/q;;;2*+1/p-2
InChI key
ATUUSMBCJWYMEG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Copper(I) triflate toluene complex is a copper-based catalyst for cross coupling reactions.[1]
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
170.0 °F - closed cup
Flash Point(C)
76.67 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service