510696
Niobium(V) chloride
anhydrous, powder, 99.995% trace metals basis
Synonym(s):
Niobium pentachloride
About This Item
Recommended Products
grade
anhydrous
Quality Level
Assay
99.995% trace metals basis
form
powder
impurities
≤55.0 ppm Trace Metal Analysis
bp
254 °C (lit.)
mp
204.7 °C (lit.)
density
2.75 g/mL at 25 °C (lit.)
SMILES string
Cl[Nb](Cl)(Cl)(Cl)Cl
InChI
1S/5ClH.Nb/h5*1H;/q;;;;;+5/p-5
InChI key
YHBDIEWMOMLKOO-UHFFFAOYSA-I
Looking for similar products? Visit Product Comparison Guide
Application
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service