Skip to Content
Merck
All Photos(2)

Key Documents

496871

Sigma-Aldrich

1,3-Bis(tert-butoxycarbonyl)guanidine

98%

Synonym(s):

C,C′-Bis(1,1-dimethylethyl) N,N′-(carbonimidoyl)bis[carbamate], Di-Boc-guanidine, N,N′-Bis(Boc)guanidine, N,N′-Di-Boc-guanidine, N1,N2-Bis(tert-butoxycarbonyl)guanidine, N1,N2-Bis(tert-butyloxycarbonyl)guanidine, tert-Butyl [N-(tert-butoxycarbonyl)carbamimidoyl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN=C[NHCO2C(CH3)3]2
CAS Number:
Molecular Weight:
259.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

141-144 °C (lit.)

SMILES string

CC(C)(C)OC(=O)NC(=N)NC(=O)OC(C)(C)C

InChI

1S/C11H21N3O4/c1-10(2,3)17-8(15)13-7(12)14-9(16)18-11(4,5)6/h1-6H3,(H3,12,13,14,15,16)

InChI key

VPWFNCFRPQFWGS-UHFFFAOYSA-N

General description

1,3-Bis(tert-butoxycarbonyl)guanidine is a guanidine derivative. The synthesis of N-substituted 2-aminoimidazole inhibitors using 1,3-bis(tert-butoxycarbonyl)guanidine has been reported.

Application

1,3-Bis(tert-butoxycarbonyl)guanidine may be used to produce a guanidinyl derivative during the multi-step synthesis of (-)-crispine E. It may also be used as a reagent during the preparation of 1,6-diphenylnaphthalenes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongzheng Zhang et al.
Bioorganic & medicinal chemistry letters, 23(7), 2001-2006 (2013-03-14)
Bacterial cell division occurs in conjunction with the formation of a cytokinetic Z-ring structure comprised of FtsZ subunits. Agents that disrupt Z-ring formation have the potential, through this unique mechanism, to be effective against several of the newly emerging multidrug-resistant
Synthesis of (-)-Trolline,(-)-Crispin A and (-)-Crispine E.
Kanemitsu T, et al.
Heterocycles, 74, 199-204 (2008)
Andrew A Yeagley et al.
Organic & biomolecular chemistry, 11(1), 130-137 (2012-10-19)
Antibiotic resistance is a significant problem and is compounded by the ability of many pathogenic bacteria to form biofilms. A library of N-substituted derivatives of a previously reported 2-aminoimidazole/triazole (2-AIT) biofilm modulator was constructed via α-bromoketone cyclization with 1,3-bis(tert-butoxycarbonyl)guanidine, followed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service