Skip to Content
Merck
All Photos(1)

Key Documents

488321

Sigma-Aldrich

Butane-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH213CH3
CAS Number:
Molecular Weight:
59.11
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

vapor density

2.11 (vs air)

vapor pressure

51.6 psi ( 37.7 °C)

isotopic purity

99 atom % 13C

autoignition temp.

761 °F

bp

−0.5 °C (lit.)

mp

−138 °C (lit.)

mass shift

M+1

SMILES string

CCC[13CH3]

InChI

1S/C4H10/c1-3-4-2/h3-4H2,1-2H3/i1+1

InChI key

IJDNQMDRQITEOD-OUBTZVSYSA-N

Related Categories

Packaging

Standard 100 mL quantitiy is packaged in a 25 mL Sure/Pac cylinder with brass 1/4" male NPT valve. Nominal gas pressure at 21ºC is 16 psig for 100 mL. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Legal Information

Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service