Skip to Content
Merck
All Photos(3)

Key Documents

480762

Sigma-Aldrich

Zinc citrate dihydrate

97%

Synonym(s):

Citric acid zinc salt, Zinc citrate tribasic dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O7)2Zn3 · 2H2O
CAS Number:
Molecular Weight:
610.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
97%
form:
powder
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

mp

334 °C (dec.) (lit.)

SMILES string

O.O.[Zn++].[Zn++].[Zn++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/2C6H8O7.2H2O.3Zn/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*1H2;;;/q;;;;3*+2/p-6

InChI key

OXAGUGIXGVHDGD-UHFFFAOYSA-H

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

  1. Is it cell culture compatible?

    1 answer
    1. This specific product has not been assayed for suitability in cell culture. There are alternative zinc salts in our catalog that have been listed as being suitable for cell culture, such as items Z0251 or Z0152.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service