Recommended Products
Assay
97%
refractive index
n20/D 1.443 (lit.)
bp
171-172 °C (lit.)
density
0.836 g/mL at 25 °C (lit.)
SMILES string
CCCCNCCN
InChI
1S/C6H16N2/c1-2-3-5-8-6-4-7/h8H,2-7H2,1H3
InChI key
DFPGBRPWDZFIPP-UHFFFAOYSA-N
General description
N-Butylethylenediamine is a chelating amine ligand.
Application
N-Butylethylenediamine may be used as a ligand in the synthesis of one dimensional copper-azido molecular tape, [Cu2(N-Buen)(N3)4]n where N-Buen = N-butylethylenediamine.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
138.2 °F - closed cup
Flash Point(C)
59 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Novel copper-azido magnetic molecular tapes: syntheses, structures, and magnetic properties.
CrystEngComm, 13(21), 6415-6421 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service