Skip to Content
Merck
All Photos(1)

Key Documents

47587

Sigma-Aldrich

Fmoc-β-Ala-OH

≥99.0% (HPLC)

Synonym(s):

Fmoc-β-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO4
CAS Number:
Molecular Weight:
311.33
Beilstein:
2302327
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (HPLC)

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

142-147 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)CCNC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C18H17NO4/c20-17(21)9-10-19-18(22)23-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16H,9-11H2,(H,19,22)(H,20,21)

InChI key

LINBWYYLPWJQHE-UHFFFAOYSA-N

General description

Fmoc-β-Ala-OH also known as Fmoc-β-alanine, is a versatile reagent for solid phase peptide synthesis.

Application

Fmoc-β-Ala-OH is used as a linker to synthesize peptides on a monolithic GMA-EDMA (glycidyl methacrylate-co-ethylene dimethacrylate) matrix via Fmoc solid phase peptide synthesis. [1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solid phase peptide synthesis on epoxy-bearing methacrylate monoliths
E Vlakh
Journal of Peptide Science, 10, 719-730 (2004)
Sofie Trier et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 96, 329-337 (2015-09-09)
Acylation of peptide drugs with fatty acid chains has proven beneficial for prolonging systemic circulation, as well as increasing enzymatic stability and interactions with lipid cell membranes. Thus, acylation offers several potential benefits for oral delivery of therapeutic peptides, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service