Skip to Content
Merck
All Photos(1)

Key Documents

464775

Sigma-Aldrich

Potassium pentachloronitrosylruthenate(II)

Synonym(s):

Dipotassium pentachloro(nitrosyl)ruthenate(II), di-Potassium nitrosylpentachlororuthenate(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Ru(NO)Cl5
CAS Number:
Molecular Weight:
386.54
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

crystalline

concentration

25.2-30.7% Ru (after reduction with magnesium, gravimetric)

mp

220 °C (dec.) (lit.)

SMILES string

[K+].[K+].Cl[Ru--](Cl)(Cl)(Cl)(Cl)N=O

InChI

1S/5ClH.2K.NO.Ru/c;;;;;;;1-2;/h5*1H;;;;/q;;;;;2*+1;-1;+4/p-5

InChI key

WNVHAHDPZULZJL-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Istvan Szundi et al.
Biophysical journal, 118(2), 386-395 (2019-12-25)
Earlier CO flow-flash experiments on the fully reduced Thermus thermophilus ba3 (Tt ba3) cytochrome oxidase revealed that O2 binding was slowed down by a factor of 10 in the presence of CO (Szundi et al., 2010, PNAS 107, 21010-21015). The goal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service