Skip to Content
Merck
All Photos(1)

Key Documents

453021

Sigma-Aldrich

Allyl alcohol

≥98.5%

Synonym(s):

2-Propen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OH
CAS Number:
Molecular Weight:
58.08
Beilstein:
605307
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2 (vs air)

Quality Level

vapor pressure

23.8 mmHg ( 25 °C)

Assay

≥98.5%

autoignition temp.

712 °F

expl. lim.

18 %

refractive index

n20/D 1.412 (lit.)

bp

96-98 °C (lit.)

mp

−129 °C (lit.)

density

0.854 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC=C

InChI

1S/C3H6O/c1-2-3-4/h2,4H,1,3H2

InChI key

XXROGKLTLUQVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Used to induce a mouse model of liver damage that has been used to study the mechanisms of hepatotoxicity and hepatic stem cell-mediated repair.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad A Khosa et al.
Journal of hazardous materials, 278, 360-371 (2014-07-06)
Chemical modification of chicken feathers (CF) and their subsequent role in arsenic removal from water is presented in this paper. The ground CF were chemically treated with four selective dopants such as poly (ethylene glycol) (PEG) diglycidyl ether, poly (N-isopropylacrylamide)
Hidenori Kinoshita et al.
Organic letters, 6(22), 4085-4088 (2004-10-22)
[reaction: see text] An aqueous biphasic reaction system enables the direct use of allyl alcohol in the Tsuji-Trost reaction without the help of any activating reagents for allyl alcohol. The reaction conditions are neutral to basic, allowing the use of
Orla M Wilson et al.
Journal of the American Chemical Society, 128(14), 4510-4511 (2006-04-06)
We report a particle size dependence for the rate of hydrogenation of allyl alcohol using 1.3-1.9 nm Pd dendrimer-encapsulated nanoparticle (DEN) catalysts. For particles with diameters of <1.5 nm and containing <147 Pd atoms, the modulation in catalytic activity is
Hydrosilylation of allyl alcohol with [HSiMe2OSiO1.5]8: octa (3-hydroxypropyldimethylsiloxy) octasilsesquioxane and its octamethacrylate derivative as potential precursors to hybrid nanocomposites.
Zhang C and Laine RM.
Journal of the American Chemical Society, 122(29), 6979-6988 (2000)
Deoxydehydration of glycerol to allyl alcohol catalyzed by rhenium derivatives.
Canale V, et al.
Catalysis Science & Technology, 4(10), 3697-3704 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service