Skip to Content
Merck
All Photos(1)

Key Documents

448206

Sigma-Aldrich

p-Cresol-d8

≥98 atom % D, ≥98% (CP)

Synonym(s):

4-Methylphenol-d8

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3C6D4OD
CAS Number:
Molecular Weight:
116.19
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥98 atom % D

Assay

≥98% (CP)
99% (CP)

refractive index

n20/D 1.537 (lit.)

bp

202 °C (lit.)

mp

32-34 °C (lit.)

density

1.126 g/mL at 25 °C (lit.)

application(s)

environmental

mass shift

M+8

SMILES string

[2H]Oc1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])[2H]

InChI

1S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3/i1D3,2D,3D,4D,5D/hD

InChI key

IWDCLRJOBJJRNH-IWRLGKISSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natsuki Ikematsu et al.
Legal medicine (Tokyo, Japan), 34, 27-35 (2018-08-14)
In some forensic autopsy cases there are high concentrations of p-cresol in the blood. In vivo, p-cresol is the only isomer yielded by intestinal bacteria and is excreted into urine. We investigated the diagnostic meaning of p-cresol in the blood
Martin Oppegård et al.
Rapid communications in mass spectrometry : RCM, 34(24), e8950-e8950 (2020-09-19)
Produced water (PW) discharge from the oil and gas industry represents the largest intentional marine waste volume. Alkyl phenols (APs) are one of the main toxic component groups found in PW, with concentration of APs in discharged PW from the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service