Skip to Content
Merck
All Photos(1)

Key Documents

440507

Sigma-Aldrich

N-(2-Hydroxyethyl)trifluoroacetamide

97%

Synonym(s):

N-(Trifluoroacetyl)ethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CONHCH2CH2OH
CAS Number:
Molecular Weight:
157.09
Beilstein:
1934914
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reagent type: cross-linking reagent

mp

32-35 °C (lit.)

density

1.413 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

O=C(NCCO)C(F)(F)F

InChI

1S/C4H6F3NO2/c5-4(6,7)3(10)8-1-2-9/h9H,1-2H2,(H,8,10)

InChI key

NPSVCXHBJVBBAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the development of a microwave-mediated method for O-allylation of amido alcohols on chromatographic alumina.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service