The molecular weight of this material is not determined on a lot to lot basis. However, historical data indicate an expected molecular weight of 2490 g/mol.
438146
Poly(propylene glycol) monobutyl ether
Synonym(s):
Butoxypolypropylene glycol, PPG-14 Butyl Ether, PPG-16 Butyl Ether, Poly(propylene oxide) monobutyl ether, Polyoxypropylene glycol butyl monoether
About This Item
Recommended Products
form
viscous liquid
refractive index
n20/D 1.45
viscosity
1300 cSt(20 °C)(lit.)
bp
>200 °C (lit.)
transition temp
pour point −23 °C
density
1 g/mL at 25 °C
SMILES string
CCCCO.CC(O)CO
InChI
1S/C10H22O3/c1-4-5-6-12-8-10(3)13-7-9(2)11/h9-11H,4-8H2,1-3H3
InChI key
CUVLMZNMSPJDON-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
285.1 °F - closed cup
Flash Point(C)
140.6 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the molecular weight of this product? The other PPG-Butyl Ether you sell is listed as Mn ~ 1,000 but this one is unlisted
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service