If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
427160
Poly(vinylidene fluoride-co-hexafluoropropylene)
average Mw ~400,000, average Mn ~130,000, pellets
Synonym(s):
PVDF-HFP
About This Item
Recommended Products
form
pellets
melt index
3.5-7.5 g/10 min (230°C/12.5kg)
mol wt
average Mn ~130,000
average Mw ~400,000
impact strength
12-20 ft-lb/in. (Izod, ASTM D 256, notched)
dielectric constant
9.4-10.6, 100 Hz (ASTM D 150)
hardness
65-70 (Shore D, ASTM D 2240)
refractive index
n20/D 1.41
viscosity
23,000-27,000 poise(230 °C) (100 sec-1; typical)(lit.)
transition temp
brittleness temperature -62 °C (ASTM 2800)
Tm 140-145 °C (ASTM D 3418)
density
1.77 g/mL at 25 °C
SMILES string
FC(F)=C.FC(F)=C(F)C(F)(F)F
InChI key
OQMIRQSWHKCKNJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
It can be used to prepare superhydrophobic polymer nanocomposites. For example, PVDF-HFP- SiO2 composite can be used to enhance the thermochromism in regioregular poly(3-hexylthiophene), P3HT.
It can also be used to prepare lead-free piezoelectric flexible polymer composites for nanogenerators. PVDF-HFP can be preferred over other polymer matrices because of its higher solubility, greater free volume, and better mechanical strength.
Features and Benefits
Physical form
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.
Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.
Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.
Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service