Skip to Content
Merck
All Photos(1)

Key Documents

419613

Sigma-Aldrich

2-Methyl-1-propenylmagnesium bromide solution

0.5 M in THF

Synonym(s):

(2-Methylpropenyl)magnesium bromide, 2,2-Dimethylvinylmagnesium bromide, Isobutenylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHMgBr
CAS Number:
Molecular Weight:
159.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

density

0.952 g/mL at 25 °C

SMILES string

C\C(C)=C\[Mg]Br

InChI

1S/C4H7.BrH.Mg/c1-4(2)3;;/h1H,2-3H3;1H;/q;;+1/p-1

InChI key

BKVIQODYSREJQH-UHFFFAOYSA-M

General description

2-Methyl-1-propenylmagnesiumbromide is a heterocyclic organic compound. It is used as a Grignard reagent in chemical synthesis.[1]

Application

Used in a study of a palladium-catalyzed coupling with aryl tosylates.[2]
Used in a synthesis of (2S,3S)-3′-fluoroisoleucine.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Oxy-Heck--Matsuda Arylations: Expeditious Synthesis of Dihydrobenzofuran Systems and Total Synthesis of the Neolignan (-)-Conocarpan
Silva AR, et al.
advanced synthesis and catalysis, 360(2), 346-365 (2018)
Jean-Damien Charrier et al.
Organic & biomolecular chemistry, 2(5), 797-802 (2004-02-27)
The fluorinated amino acid, (2S,3S)-3'-fluoroisoleucine 3, has been synthesised by a route involving stereoselective cuprate or photochemical addition to the compound 4, derived in turn from (2S)-pyroglutamic acid. This provides a reporter group for the hydrophobic amino acid (2S)-isoleucine for
Michael E Limmert et al.
The Journal of organic chemistry, 70(23), 9364-9370 (2005-11-05)
[Reaction: see text]. Aryl and alkenyl tosylates are easily prepared, inexpensive and, thus, attractive for transition-metal-catalyzed couplings, but their reactivity is low. We report examples of mild, palladium-catalyzed coupling of aryl, alkenyl, and alkyl Grignard reagents with aryl and alkenyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service