Skip to Content
Merck
All Photos(2)

Key Documents

419184

Sigma-Aldrich

Triphenylphosphine diiodide

technical grade, 90%

Synonym(s):

Diiodotriphenylphosphorane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3PI2
CAS Number:
Molecular Weight:
516.09
Beilstein:
2740417
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.06

grade

technical grade

Assay

90%

form

solid

mp

210-220 °C (lit.)

functional group

phosphine

SMILES string

IP(I)(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15I2P/c19-21(20,16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

NSWZEXJQHIXCFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triphenylphosphine diiodide (Ph3P. I2) can be used as a reagent:
  • To facilitate the conversion of alcohol, thiol, and enol functional groups into alkyl iodides via formation of alkoxyphosphonium iodide.
  • To prepare β-iodo-α,β-unsaturated ketones from cyclic β-diketones in the presence of triethyl amine.
  • In the synthesis of alkyl nitrates from alcohols.
  • In the Beckmann rearrangement reaction to synthesize lactams from cycloalkanone oxime.
  • For the preparation of aromatic and aliphatic carboxylic acid esters in the presence of N,N-dimethylaminopyridine.

Ph3P. I2 can also be used to reduce graphene oxide for the production of graphene nanosheets under mild and environmentally friendly conditions.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New approach for the reduction of graphene oxide with triphenylphosphine dihalide
Shin H-S et al.
Royal Society of Chemistry Advances, 6(23), 18809-18813 (2016)
One-Pot Synthesis of Carboxylic Acid Esters in Neutral and Mild Conditions by Triphenylphosphine Dihalide [Ph 3 PX 2 (X= Br, I)].
Sardarian Alireza R, et al.
Acta Chimica Slovenica, 56(3) (2009)
Triphenylphosphine-Iodine
Dormoy J-R
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service