Skip to Content
Merck
All Photos(2)

Key Documents

418870

Sigma-Aldrich

Poly(acrylonitrile-co-butadiene), dicarboxy terminated

average Mn ~3,800, acrylonitrile 8-12 wt. %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C[CH2CH(CN)]x(CH2CH=CHCH2)yCO2H
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

viscous liquid

specific activity

1.9 carboxyl groups per molecule

mol wt

average Mn ~3,800

composition

acrylonitrile, 8-12 wt. %

refractive index

n20/D 1.515 (lit.)

viscosity

600 poise(27 °C, Brookfield)(lit.)

acid number

28

transition temp

Tg −66 °C (DSC)

density

0.924 g/mL at 25 °C

SMILES string

N#CC=C.C(=C)C=C

InChI

1S/C4H6.C3H3N/c1-3-4-2;1-2-3-4/h3-4H,1-2H2;2H,1H2

InChI key

NTXGQCSETZTARF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Component of epoxy-based adhesives, composites and coatings. Intermediate in corrosion-resistant vinyl esters, acrylated epoxies used in radiation curing and photopolymer printing plates.

Features and Benefits

Enhances impact strength, thermal shock, peel strength and crack resistance of epoxy compositions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service