Skip to Content
Merck
All Photos(1)

Key Documents

407364

Sigma-Aldrich

2-Hydroxy-3-phenoxypropyl acrylate

contains 250 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

(2-Hydroxy-3-phenoxypropyl) prop-2-enoate, 3-Phenoxy-2-hydroxypropyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CH(OH)CH2OC6H5
CAS Number:
Molecular Weight:
222.24
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid

contains

250 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.528 (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

OC(COC(=O)C=C)COc1ccccc1

InChI

1S/C12H14O4/c1-2-12(14)16-9-10(13)8-15-11-6-4-3-5-7-11/h2-7,10,13H,1,8-9H2

InChI key

HHQAGBQXOWLTLL-UHFFFAOYSA-N

Application

Polymeric membranes for controlled drug release systems may be prepared by the photosynthesis of 2-hydroxy-3-phenoxypropyl acrylate by UV radiation. [1] Tthe product can be used as a curing agent during the fabrication of poly (phenylene ether)-based substrate materials. [2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

345.2 °F - closed cup

Flash Point(C)

174 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A brief view on antihypertensive drugs delivery through transdermal patches.
Rastogi V, et al.
International Journal of Pharmaceutical Sciences and Research, 3(7) (2012)
Synthesis and Analysis of Multi-functional Urethane Acrylate Monomer, and its Application as Curing Agent for Poly (phenylene ether)-based Substrate Material.
Kim DK, et al.
Polymer (Korea), 36(4), 413-419 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service