Skip to Content
Merck
All Photos(2)

Key Documents

400777

Sigma-Aldrich

Nickel(II) iodide

powder

Synonym(s):

Nickel diiodide, Nickel iodide, Nickelous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiI2
CAS Number:
Molecular Weight:
312.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

73.1-89.0% iodine (I) basis (silver nitrate titration)

form

powder

reaction suitability

reagent type: catalyst
core: nickel

mp

797 °C (lit.)

density

5.83 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

I[Ni]I

InChI

1S/2HI.Ni/h2*1H;/q;;+2/p-2

InChI key

BFSQJYRFLQUZKX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Machrouhi, F. Namy, J-L.
Tetrahedron Letters, 40, 1315-1315 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service