Skip to Content
Merck
All Photos(1)

Key Documents

343870

Sigma-Aldrich

1-Bromo-3-(trifluoromethoxy)benzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OCF3
CAS Number:
Molecular Weight:
241.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.462 (lit.)

density

1.62 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

FC(F)(F)Oc1cccc(Br)c1

InChI

1S/C7H4BrF3O/c8-5-2-1-3-6(4-5)12-7(9,10)11/h1-4H

InChI key

WVUDHWBCPSXAFN-UHFFFAOYSA-N

General description

1-Bromo-3-(trifluoromethoxy)benzene undergoes Diels-Alder reaction with lithium diisopropylamide (LDA) in THF and furan, to yield the corresponding 1,4-dihydro-1,4-epoxy-5- or 6-(trifluoromethoxy)naphthalenes.

Application

1-Bromo-3-(trifluoromethoxy)benzene may be used in the preparation of 1,2-dehydro-3-(trifluoromethoxy)benzene. It may be used in the synthesis of new electronically deficient atropisomeric diphosphine ligand (S)-CF3O-BiPhep [2,2′-bis(diphenylphosphino)-6,6′-ditrifluoromethoxy-1,1′-biphenyl].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 2-Didehydro-3-and-4-(trifluoromethoxy) benzene: The ?Aryne? Route to 1-and 2-(Trifluoromethoxy) naphthalenes.
Schlosser M and Castagnetti E.
European Journal of Organic Chemistry, 21, 3991-3997 (2001)
The use of arynes in organic synthesis.
Pellissier H and Santelli M.
Tetrahedron, 59(6), 701-730 (2003)
A new electronically deficient atropisomeric diphosphine ligand (S)-CF3O-BiPhep and its application in asymmetric hydrogenation.
Zhang D-Y, et al.
Tetrahedron Letters, 53(20), 2556-2559 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service