Skip to Content
Merck
All Photos(1)

Key Documents

341800

Sigma-Aldrich

1,4,4a,8a-Tetrahydro-endo-1,4-methanonaphthalene-5,8-dione

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O2
CAS Number:
Molecular Weight:
174.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

77-79 °C (lit.)

SMILES string

O=C1C=CC(=O)[C@H]2[C@@H]3C[C@@H](C=C3)[C@@H]12

InChI

1S/C11H10O2/c12-8-3-4-9(13)11-7-2-1-6(5-7)10(8)11/h1-4,6-7,10-11H,5H2/t6-,7+,10+,11-

InChI key

FQLRTGXTYFCECH-FIPCFZRWSA-N

General description

1,4,4a,8a-Tetrahydro-endo-1,4-methanonaphthalene-5,8-dione can be prepared by Diels-Alder reaction of cyclopentadiene and 1,4-benzoquinone.

Application

1,4,4a,8a-Tetrahydro-endo-1,4-methanonaphthalene-5,8-dione may be used in the synthesis of pentacyclo[5.4.0.02,6.03,10.05,9]undecane-8,11-dione (the ′cage′ compound) via the intramolecular [2+2]-photocycloaddition reaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical synthesis of a ?cage? compound in a microreactor: Rigorous comparison with a batch photoreactor.
Aillet T, et al.
Chemical Engineering and Processing, 64, 38-47 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service