Skip to Content
Merck
All Photos(2)

Documents

324922

Sigma-Aldrich

(−)-Methyl (S)-3-bromo-2-methylpropionate

97%

Synonym(s):

(−)-Methyl (S)-β-bromoisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH(CH3)CO2CH3
CAS Number:
Molecular Weight:
181.03
Beilstein:
1720832
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]20/D −23 to −16°, c = 5 neat

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.455 (lit.)

bp

97.5-98.5 °C/62 mmHg (lit.)

density

1.422 g/mL at 25 °C (lit.)

SMILES string

COC(=O)[C@H](C)CBr

InChI

1S/C5H9BrO2/c1-4(3-6)5(7)8-2/h4H,3H2,1-2H3/t4-/m1/s1

InChI key

FKWNAVCXZSQYTA-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(−)-Methyl (S)-3-bromo-2-methylpropionate can be used:
  • As a reference substrate in the alkylation reactions of vitamin B12 derivatives.
  • As a starting material in the preparation of biologically significant 3-[(2H-1,2,4-benzothiadiazine-1,1-dioxide-3-yl)thio]-2-methylpropanoic acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereospecific synthesis of 3-[(2H-1, 2, 4-Benzothiadiazine-1, 1-dioxide-3-yl) thio]-2-methylpropanoic acids
Tait A, et al.
Tetrahedron Asymmetry, 7(9), 2703-2706 (1996)
Hydrophobic vitamin B12. Part 12. Preparation, characterization and enantioselective alkylation of strapped hydrophobic-vitamin B12
Murakami Y, et al.
J. Chem. Soc. Perkin Trans. II, 1175-1183 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service