Skip to Content
Merck
All Photos(2)

Key Documents

311200

Sigma-Aldrich

4-Methyl-3-penten-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2OH
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

157 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C\C(C)=C\CCO

InChI

1S/C6H12O/c1-6(2)4-3-5-7/h4,7H,3,5H2,1-2H3

InChI key

FKKLUOCEIANSFL-UHFFFAOYSA-N

Application

4-Methyl-3-penten-1-ol was used in the synthesis of tetrahydrofuro[3,2-c]benzothiopyran with a trans:cis ratio of 78:22.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile and efficient one-pot synthesis of thiochromans from bis (2-formylphenyl) disulfide and alkenols via iodine-promoted generation and subsequent intramolecular cycloaddition of< i> ortho</i>-thiobenzoquinone methides.
Tetrahedron Letters, 44(34), 6513-6517 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service