Skip to Content
Merck
All Photos(2)

Key Documents

303348

Sigma-Aldrich

Cyclopentadienyllithium

97%

Synonym(s):

1,3-Cyclopentadiene lithium complex, Lithium cyclopentadienide, Lithium cyclopentadienylide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Li
CAS Number:
Molecular Weight:
72.03
Beilstein:
3535737
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

reaction suitability

reagent type: ligand

SMILES string

[Li]C1C=CC=C1

InChI

1S/C5H5.Li/c1-2-4-5-3-1;/h1-5H;

InChI key

JTVVTAXHEPEOQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful reagent for the preparation of metallocenes and cyclopentadienyl derivatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service