Skip to Content
Merck
All Photos(1)

Key Documents

301477

Sigma-Aldrich

Tri(ethylene glycol) bis(chloroformate)

96%

Synonym(s):

Triethylene glycol bischloroformate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCO2CH2CH2OCH2)2
CAS Number:
Molecular Weight:
275.08
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

0.09 psi ( 20 °C)

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.457 (lit.)

density

1.34 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)OCCOCCOCCOC(Cl)=O

InChI

1S/C8H12Cl2O6/c9-7(11)15-5-3-13-1-2-14-4-6-16-8(10)12/h1-6H2

InChI key

IFOIGJKHVZBFPR-UHFFFAOYSA-N

Application


  • Tailoring the Surface Chemistry of PEDOT: PSS to Promote Supported Lipid Bilayer Formation: Details the method of using tri(ethylene glycol) bis(chloroformate) in surface chemistry applications (K Kallitsis et al., 2023).

  • Combinations of dual-function azide-containing crosslinkers with CH insertion capabilities for polymeric elastomers with improved adhesion properties: Discusses the preparation and application of tri(ethylene glycol)diazidoformate starting from tri(ethylene glycol)bis(chloroformate) (DW Jung et al., 2022).

  • Preparation of polypropylene with strain hardening characteristics via the noncatalytic C-H insertion of azidoformate: Explores the reaction process involving tri(ethylene glycol) bis(chloroformate) (SY Kim et al., 2022).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service