Skip to Content
Merck
All Photos(2)

Key Documents

279773

Sigma-Aldrich

1-(4-Nitrophenyl)piperazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3O2
CAS Number:
Molecular Weight:
207.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

mp

131-133 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1ccc(cc1)N2CCNCC2

InChI

1S/C10H13N3O2/c14-13(15)10-3-1-9(2-4-10)12-7-5-11-6-8-12/h1-4,11H,5-8H2

InChI key

VWOJSRICSKDKAW-UHFFFAOYSA-N

General description

Inclusion interactions of 1-(4-nitrophenyl)piperazine with 4-sulfonatocalix[n]arenes were investigated by UV spectroscopy and fluorescence spectroscopy[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongbin Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 132, 44-51 (2014-05-27)
The inclusion behaviors of 4-Sulfonatocalix[n]arenes (SCXn) (n=4, 6, 8) with 1-(4-nitrophenyl)piperazine (NPP) were investigated by UV spectroscopy and fluorescence spectroscopy at different pH values (pH=3.05, 6.50, 8.40). The UV absorption and fluorescence intensity of NPP remarkably increased in presence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service