Skip to Content
Merck
All Photos(1)

Key Documents

267848

Sigma-Aldrich

3-Bromopentane

95%

Synonym(s):

3-Pentyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2CHBr
CAS Number:
Molecular Weight:
151.04
Beilstein:
1730967
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

refractive index

n20/D 1.445 (lit.)

bp

118-119 °C (lit.)

density

1.216 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CCC(Br)CC

InChI

1S/C5H11Br/c1-3-5(6)4-2/h5H,3-4H2,1-2H3

InChI key

VTOQFOCYBTVOJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Bromopentane is a glass forming liquid. The dielectric relaxation of 3-bromopentane was studied[1].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Huang et al.
The Journal of chemical physics, 123(16), 164504-164504 (2005-11-05)
The glass-forming liquids 3-bromopentane (3BP) and 3-methylpentane (3MP) are readily miscible across the entire composition range, although their polarities differ considerably. As noted by Berberian [J. Non-Cryst. Solids 131-133, 48 (1991)], the nearly matching molar volumes makes this binary system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service