Skip to Content
Merck
All Photos(1)

Key Documents

264768

Sigma-Aldrich

Furfuryl disulfide

95%

Synonym(s):

Difurfuryl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2S2
CAS Number:
Molecular Weight:
226.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.585 (lit.)

bp

112-115 °C/0.5 mmHg (lit.)

mp

10-11 °C (lit.)

density

1.233 g/mL at 25 °C (lit.)

functional group

disulfide

SMILES string

C(SSCc1ccco1)c2ccco2

InChI

1S/C10H10O2S2/c1-3-9(11-5-1)7-13-14-8-10-4-2-6-12-10/h1-6H,7-8H2

InChI key

CBJPZHSWLMJQRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Furfuryl disulfide has been characterized as an important aroma constituent of roasted coffee[1].

Application

Furfuryl disulfide has been used in the preparation of poly(disulfido-imide)s by Diels-Alder (DA) reaction with the various bismaleimides[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Poly (imide) s based on furfuryl-disulfide.
Patel HS and Patel HD.
Phosphorus, Sulfur, and Silicon and the Related Elements, 79(1-4), 69-76 (1993)
Investigation of sulfur-containing components in roasted coffee.
R Tressl et al.
Journal of agricultural and food chemistry, 29(5), 1078-1082 (1981-09-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service