262412
4-(Butylamino)benzoic acid
97%, for peptide synthesis
Synonym(s):
p-Butylaminobenzoic acid
About This Item
Product Name
4-(Butylamino)benzoic acid, 97%
Quality Level
Assay
97%
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
mp
149-151 °C (lit.)
application(s)
peptide synthesis
SMILES string
CCCCNc1ccc(cc1)C(O)=O
InChI
1S/C11H15NO2/c1-2-3-8-12-10-6-4-9(5-7-10)11(13)14/h4-7,12H,2-3,8H2,1H3,(H,13,14)
InChI key
YCCRFDDXAVMSLM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service