Skip to Content
Merck
All Photos(1)

Documents

259780

Sigma-Aldrich

2,4,6-Trimethylstyrene

95%, contains <0.05% tert-butylcatechol as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CH=CH2
CAS Number:
Molecular Weight:
146.23
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

liquid

contains

<0.05% tert-butylcatechol as inhibitor

refractive index

n20/D 1.532 (lit.)

bp

209 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(C=C)c(C)c1

InChI

1S/C11H14/c1-5-11-9(3)6-8(2)7-10(11)4/h5-7H,1H2,2-4H3

InChI key

PDELBHCVXBSVPJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service