Skip to Content
Merck
All Photos(1)

Key Documents

253839

Sigma-Aldrich

3-Acetylbenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC6H4CO2H
CAS Number:
Molecular Weight:
164.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

mp

169-171 °C (lit.)

functional group

carboxylic acid
ketone

SMILES string

CC(=O)c1cccc(c1)C(O)=O

InChI

1S/C9H8O3/c1-6(10)7-3-2-4-8(5-7)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

CHZPJUSFUDUEMZ-UHFFFAOYSA-N

Application

3-Acetylbenzoic acid has been used in preparation of 3-(2-hydroxyethyl)benzyl alcohol[1] and methyl 3-acetylbenzoate[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincenzo Calderone et al.
The Journal of pharmacy and pharmacology, 60(2), 189-195 (2008-02-02)
This work describes the synthesis of some benzoic (1-4) and alcoholic (5-7) nitrooxy derivatives, which are nitric oxide (NO) donors in themselves, and can also be seen as useful linkers that can be used in multi-target drugs capable of releasing
Yanong D Wang et al.
Bioorganic & medicinal chemistry, 17(5), 2091-2100 (2009-02-10)
Checkpoint deficiency of malignant cells can be exploited in cancer drug discovery. Compounds that selectively kill checkpoint-deficient cells versus checkpoint-proficient cells can be utilized to preferentially target tumor cells, while sparing normal cells. The protein p21(Wafl/Cipl/Sdi1) (hereafter referred to as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service