Skip to Content
Merck
All Photos(2)

Key Documents

251976

Sigma-Aldrich

2,4-Dichloro-1-naphthol

95%

Synonym(s):

2,4-Dichloro-α-naphthol, 2,4-Dichloro-1-naphthalenol, 2,4-Dichloronaphthol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C10H5OH
CAS Number:
Molecular Weight:
213.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
95%

vapor density

7.3 (vs air)

Quality Level

Assay

95%

form

solid

mp

104-107 °C (lit.)

solubility

pyridine: soluble 5%, clear, colorless to dark yellow

functional group

chloro

SMILES string

Oc1c(Cl)cc(Cl)c2ccccc12

InChI

1S/C10H6Cl2O/c11-8-5-9(12)10(13)7-4-2-1-3-6(7)8/h1-5,13H

InChI key

HVLJEMXDXOTWLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dichloro-1-naphthol has been used in:
  • spectrophotometric determination of arginine with previous modifications of the Sakaguchi reaction
  • guanidinoacetate and guanidinosuccinate assay of urine, an indicator of kidney dysfunction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urinary guanidinoacetate-guanidinosuccinate ratio: an indicator of kidney dysfunction.
M Sasaki et al.
Clinical chemistry, 19(3), 315-321 (1973-03-01)
Modification of the Sakaguchi reaction: spectrophotometric determination of arginine in proteins without previous hydrolysis.
Messineo L.
Archives of Biochemistry and Biophysics, 117(3), 534-540 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service