Skip to Content
Merck
All Photos(1)

Key Documents

249858

Sigma-Aldrich

Antimony(V) chloride solution

1.0 M in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbCl5
CAS Number:
Molecular Weight:
299.03
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
liquid
Pricing and availability is not currently available.

form

liquid

Quality Level

concentration

1.0 M in methylene chloride

density

1.442 g/mL at 25 °C

SMILES string

Cl[Sb](Cl)(Cl)(Cl)Cl

InChI

1S/5ClH.Sb/h5*1H;/q;;;;;+5/p-5

InChI key

VMPVEPPRYRXYNP-UHFFFAOYSA-I

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Ox. Liq. 2 - Ozone 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Urogenital tract, Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flávio José Dutra de Moura et al.
Toxicology, 243(1-2), 66-74 (2007-11-21)
Tumor necrosis factor-alpha (TNF-alpha) is a mediator of inflammation and has an important role in human and experimental renal diseases. Pentoxifylline (PTX) has been shown to inhibit cytokine synthesis, including TNF-alpha. The aim of the present study was to examine
J Carrió et al.
Experimental parasitology, 95(3), 209-214 (2000-08-31)
The determination of the intrinsic sensitivity of Leishmania strains to pentavalent antimonials in clinical trials, before treatment is begun, is essential in order to avoid failures and to allow alternative drugs to be chosen. A comparative study of SbV activity
M P Oliveira-Neto et al.
Ophthalmology, 107(1), 169-172 (2000-01-27)
To describe American cutaneous leishmaniasis of the eyelids and highlight the main clinical and diagnostic features of lesions, which are rare in this location. Retrospective, noncomparative case series Leishmanin skin test, touch preparations, histopathologic analysis, and culture in appropriate media
In-transit chemical spill--West Virginia.
MMWR. Morbidity and mortality weekly report, 35(6), 81-82 (1986-02-14)
Et2SBrSbCl5Br: an effective reagent for direct bromonium-induced polyene cyclizations.
Scott A Snyder et al.
Angewandte Chemie (International ed. in English), 48(42), 7899-7903 (2009-09-17)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service