Skip to Content
Merck
All Photos(1)

Documents

225967

Sigma-Aldrich

2-Nitrobenzylamine hydrochloride

Synonym(s):

2-Nitrobenzenemethanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2NH2 · HCl
CAS Number:
Molecular Weight:
188.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

247 °C (dec.) (lit.)

SMILES string

Cl.NCc1ccccc1[N+]([O-])=O

InChI

1S/C7H8N2O2.ClH/c8-5-6-3-1-2-4-7(6)9(10)11;/h1-4H,5,8H2;1H

InChI key

BASJTVIZZDEQBJ-UHFFFAOYSA-N

Application

2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies.
Reactant involved in synthesis of:
  • Tetrahydroisoquinolines via Heck-aza-Michael reactions
  • Aminomethylphenylamine scaffolds for factor Xa inhibition
  • Squaryliamides for mannosyltransferase inhibition
  • Fluorescent two-photon absorption benzothiadiazole dyes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service