Skip to Content
Merck
All Photos(1)

Key Documents

220701

Sigma-Aldrich

L-Phenylalanine ethyl ester hydrochloride

99%

Synonym(s):

(S)-Phenylalanine ethyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)COOC2H5 · HCl
CAS Number:
Molecular Weight:
229.70
Beilstein:
3657823
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −7.8°, c = 2 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

155-156 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.CCOC(=O)[C@@H](N)Cc1ccccc1

InChI

1S/C11H15NO2.ClH/c1-2-14-11(13)10(12)8-9-6-4-3-5-7-9;/h3-7,10H,2,8,12H2,1H3;1H/t10-;/m0./s1

InChI key

FPFQPLFYTKMCHN-PPHPATTJSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas J Reichert et al.
Protein engineering, design & selection : PEDS, 28(12), 553-565 (2015-09-26)
The bioorthogonal keto group has attracted interest for the site-specific chemical conjugation of recombinant proteins under mild conditions, e.g. with aminooxy-functionalised fluorescent probes, radiometal chelates, toxins or polymers. However, the cotranslational incorporation of the corresponding non-canonical amino acid p-acetyl-L-phenylalanine (Apa)
Gayong Shim et al.
Nanotoxicology, 11(6), 762-770 (2017-07-08)
Poly-γ-glutamic acid (PGA), a major component of the bacterial capsule, is known to confer hydrophilicity to bacterial surfaces and protect bacteria from interactions with blood cells. We tested whether applying a bacteriomimetic surface coating of PGA modulates interactions of nanomaterials
João Albuquerque et al.
Scientific reports, 10(1), 6830-6830 (2020-04-24)
The amino acid requirements of high-production dairy cows represent a challenge to ensuring that their diet is supplied with available dietary resources, and thus supplementation with protected amino acids is necessary to increase their post-ruminal supply. Lysine is often the
Dongyoon Kim et al.
Biomaterials science, 8(4), 1106-1116 (2020-01-30)
In this study, we sought to design a bionanomaterial that could exert anticancer effects against primary tumors and protect against rechallenged tumors via photodynamic immunotherapy. As a biomaterial, we used an amphiphilic phenylalanine derivative of poly-gamma glutamic acid, which forms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service