Skip to Content
Merck
All Photos(1)

Key Documents

219576

Sigma-Aldrich

(1S)-(+)-10-Camphorsulfonyl chloride

97%

Synonym(s):

(+)-Camphor-10-sulfonyl chloride, (1S)-Camphor-10-sulfonic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15ClO3S
CAS Number:
Molecular Weight:
250.74
Beilstein:
3205974
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

65-67 °C (lit.)

SMILES string

[H][C@@]12CC[C@@](CS(Cl)(=O)=O)(C(=O)C1)C2(C)C

InChI

1S/C10H15ClO3S/c1-9(2)7-3-4-10(9,8(12)5-7)6-15(11,13)14/h7H,3-6H2,1-2H3/t7-,10-/m1/s1

InChI key

BGABKEVTHIJBIW-GMSGAONNSA-N

Application

Chiral resolving agent. Used in the synthesis of nonpeptide oxytocin antagonists.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service