Skip to Content
Merck
All Photos(1)

Documents

218286

Sigma-Aldrich

(+)-Dihydrocarvone, mixture of isomers

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.2 (vs air)

Quality Level

vapor pressure

0.06 mmHg ( 20 °C)

Assay

98%

form

liquid

optical activity

[α]22/D +17°, neat

refractive index

n20/D 1.471 (lit.)

bp

87-88 °C/6 mmHg (lit.)

density

0.929 g/mL at 25 °C (lit.)

SMILES string

CC1CCC(CC1=O)C(C)=C

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)10(11)6-9/h8-9H,1,4-6H2,2-3H3

InChI key

AZOCECCLWFDTAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Dihydrocarvone (mixture of isomers) is a monoterpenoid compound, mainly used as a building block to prepare sesquiterpenes.

Application

The N-functionalization of dihydrocarvone forms imine derivatives, which are synthons for preparing N-heterocycles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

N-functionalization of dihydrocarvone: Obtaining aminocyclohexane derivatives and their spectrometric study.
Kouznetsov VV and Stashenko EE.
Journal of the Chilean Chemical Society, 50(3), 559-563 (2005)
Hyo-Rim Lee et al.
Journal of economic entomology, 110(2), 552-557 (2017-02-07)
We evaluated the repellent activity of 12 Apiaceae plant essential oils and their components against male and female adult German cockroaches, Blattella germanica L., to find new natural repellents. Of all the plant essential oils tested, ajowan (Trachyspermum ammi Sprague)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service