Skip to Content
Merck
All Photos(3)

Key Documents

199494

Sigma-Aldrich

1-Bromooctadecane

≥97.0%

Synonym(s):

Octadecyl bromide, Stearyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17Br
CAS Number:
Molecular Weight:
333.39
Beilstein:
774145
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

solid

bp

214-216 °C/12 mmHg (lit.)

mp

25-30 °C (lit.)

density

0.976 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCBr

InChI

1S/C18H37Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3

InChI key

WSULSMOGMLRGKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromooctadecane is a key building block that can undergo nucleophilic substitution. Used to functionalize carbon nanotubes.

Application

1-Bromooctadecane was used in the synthesis of shortened single-walled carbon nanotubes (s-SWCNTs).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functionalization of shortened SWCNTs using esterification
Wan-Joong K et al.
Bulletin of the Korean Chemical Society,, 25, 1301-1302 (2004)
Kim WJ, et al.
Bull. Korean Chem. Soc., 25(9), 1301-1302 (2004)
Md Shahruzzaman et al.
Journal of separation science, 38(14), 2403-2413 (2015-05-07)
The amphiphilic polymer-grafted silica was newly prepared as a stationary phase in high-performance liquid chromatography. Poly(4-vinylpyridine) with a trimethoxysilyl group at one end was grafted onto porous silica particles and the pyridyl side chains were quaternized with 1-bromooctadecane. The obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service