Skip to Content
Merck
All Photos(2)

Documents

156035

Sigma-Aldrich

3-(Trifluoromethyl)phenol

99%

Synonym(s):

α,α,α-Trifluoro-m-cresol, 3-Hydroxybenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4OH
CAS Number:
Molecular Weight:
162.11
Beilstein:
2045663
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.56 mmHg ( 40 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.458 (lit.)

bp

178-179 °C (lit.)

mp

−2-−1.8 °C (lit.)

density

1.333 g/mL at 25 °C (lit.)

SMILES string

Oc1cccc(c1)C(F)(F)F

InChI

1S/C7H5F3O/c8-7(9,10)5-2-1-3-6(11)4-5/h1-4,11H

InChI key

UGEJOEBBMPOJMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Trifluoromethyl)phenol (3-Hydroxybenzotrifluoride) was used in the preparation of travoprost (antiglaucoma agent).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boulton LT, et al.
Organic Process Research & Development, 6(2), 138-145 (2002)
Akira Ohashi et al.
Talanta, 146, 789-794 (2015-12-24)
The solubilities of several cobalt(III) chelates (CoL3) with in supercritical carbon dioxide (SC-CO2) were investigated in the presence of fluorine- and trifluoromethyl-substituted phenols (PhOH) using UV-vis spectrophotometry. Solubility enhancement of CoL3 complexes in SC-CO2 was accomplished by adding PhOH. The
The many roles for fluorine in medicinal chemistry.
William K Hagmann
Journal of medicinal chemistry, 51(15), 4359-4369 (2008-06-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service