Skip to Content
Merck
All Photos(1)

Key Documents

102369

Sigma-Aldrich

1,4-Dinitrobenzene

98%

Synonym(s):

p-dinitrobenzene, para-Dinitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NO2)2
CAS Number:
Molecular Weight:
168.11
Beilstein:
1105828
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

183.4 °C/34 mmHg (lit.)

mp

170-173 °C (lit.)

solubility

alcohol: soluble 1g in 300ml
boiling water: soluble 1g in 555ml
cold water: soluble 1g in 12,500ml
benzene: very slightly soluble
chloroform: very slightly soluble
ethyl acetate: very slightly soluble

density

1.625 g/mL at 25 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H4N2O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4H

InChI key

FYFDQJRXFWGIBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dinitrobenzene is commonly used in industrial explosives containing ammonium nitrate. 1,4-Dinitrobenzene forms negative molecular ion M-· by electron capture or charge exchange.

Application

1,4-Dinitrobenzene was used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

Preparation Note

One gram of 1,4-Dinitrobenzene dissolves in 12,500 ml cold water, 555 ml boiling water and 300 ml alcohol.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiina J Kauppila et al.
Journal of the American Society for Mass Spectrometry, 15(2), 203-211 (2004-02-10)
The ionization mechanism in the novel atmospheric pressure photoionization mass spectrometry (APPI-MS) in negative ion mode was studied thoroughly by the analysis of seven compounds in 17 solvent systems. The compounds possessed either gas-phase acidity or positive electron affinity, whereas
Polymer sensors for nitroaromatic explosives detection.
Toal SJ and Trogler WC.
Journal of Materials Chemistry, 16(28), 2871-2883 (2006)
Silvia M Barolo et al.
The Journal of organic chemistry, 71(22), 8493-8499 (2006-10-27)
The photostimulated intramolecular ortho-arylation reactions of bromoarenes linked with pendant phenoxy containing N-substituted tetrahydroisoquinolines in liquid ammonia afforded aporphine (54-82% yield) alkaloid derivatives via SRN1 reactions. This strategy was extended for the first time to the synthesis of a homoaporphine
N Kessler et al.
Proteins, 34(3), 383-394 (1999-02-19)
The sequences of the variable regions of three monoclonal antibodies with different specificities to cholesterol monohydrate and 1,4-dinitrobenzene crystals were determined. The structures of their binding sites were then modeled, based on homology to other antibodies of known structure. Two
M Elena Hernández-Flores et al.
Steroids, 149, 108420-108420 (2019-06-04)
β-Sitosteryl (S)-ibuprofenate (2), stigmasteryl (S)-ibuprofenate (3), ergosteryl (S)-ibuprofenate (4), and cholesteryl (S)-ibuprofenate (5) were prepared in 70-75% yields by Steglich esterification and were characterized by 1D and 2D NMR, as well as by MS. The new esters were evaluated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service