Merck
Mucosa-plate for direct evaluation of mucoadhesion of drug carriers.
International journal of pharmaceutics (2012-12-29)
Amornset Tachaprutinun, Porntip Pan-In, Supason Wanichwecharungruang
ABSTRACT
The method to prepare mucosa-plates, glass slides covalently coated with mucin, is demonstrated. The use of the plate to evaluate mucoadhesion of nanocarriers made from different four polymeric materials, N-succinylchitosan (NS-chitosan), alginate (ALG), ethylcellulose (EC), and a blend of EC and methylcellulose (EC/MC), was demonstrated. While different mucoadhesion of the four carriers could be detected using mucosa-plate, the conventional viscosity measurement could not differentiate their mucin-binding ability. ALG and NS-chitosan nanospheres showed the best attachment to the mucosa-plate compared to the EC/MC and EC spheres. Capsaicin, a model hydrophobic drug, was loaded into the carriers and the ability of the different polymeric carriers to retain capsaicin at the stomach tissue was compared using an ex vivo fresh porcine stomach assay. Ability to retain capsaicin at the stomach tissue correlated well with binding affinity toward the mucosa-plate and the loading capacity of the carriers.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl cellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl cellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl cellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl